Ferrozine mono-sodium salt hydrate

Supplier: Molekula

Synonyms: FerroZine iron reagent hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate, 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulphonic acid monosodium salt hydrate, PDT disulphonate monosodium salt hydrate

M16507877
MOLEM16507877EA 0 GBP
MOLEM16507877
Ferrozine mono-sodium salt hydrate
Ferrozine mono-sodium salt hydrate
Formula: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
MDL Number: MFCD00150794
CAS Number: 63451-29-6
EINECS: 274-196-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR