2-(Bromomethyl)naphthalene

Supplier: Molekula

45435688-25G
MOLE45435688-25GEA 0 GBP
MOLE45435688-25G
2-(Bromomethyl)naphthalene
2-(Bromomethyl)naphthalene
Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 293
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004123
CAS Number: 939-26-4
EINECS: 213-359-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR