4-Aminoantipyrine

Supplier: Molekula
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

43057307-500G
MOLE43057307-500GEA 0 GBP
MOLE43057307-500G
4-Aminoantipyrine
4-Aminoantipyrine
Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR