PIPES

Supplier: Molekula

Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid

37406078-500G 37406078-100G
MOLE37406078-500GEA 273 GBP
MOLE37406078-500G MOLE37406078-100G
PIPES
PIPES
Formula: C₈H₁₈N₂O₆S₂
MW: 302.37 g/mol
Boiling Pt: 499
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00006159
CAS Number: 5625-37-6
EINECS: 227-057-6
Merck Index: 12,07633
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR