Calcium chloride dihydrate

Supplier: Molekula
Warning

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

29778875-1KG
MOLE29778875-1KGEA 144 GBP
MOLE29778875-1KG
Calcium chloride dihydrate
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR