5-Fluoro-2'-deoxyuridine (floxuridine)

Supplier: Molekula

Synonyms: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine

27134875-1G
MOLE27134875-1GEA 71.8 GBP
MOLE27134875-1G
5-Fluoro-2'-deoxyuridine (floxuridine)
5-Fluoro-2'-deoxyuridine (Floxuridine)
Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
MDL Number: MFCD00006530
CAS Number: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR