D(-)-Luciferin sodium salt monohydrate

Supplier: Molekula

25302670-25MG
MOLE25302670-25MGEA 0 GBP
MOLE25302670-25MG
D(-)-Luciferin sodium salt monohydrate
D(-)-Luciferin sodium salt
Formula: C₁₁H₇N₂NaO₃S₂·H₂O
MW: 320.32 g/mol
MDL Number: MFCD11865368
CAS Number: 103404-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR