5-Methoxysalicylaldehyde

Supplier: Molekula

Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde

22481955-5G
MOLE22481955-5GEA 0 GBP
MOLE22481955-5G
5-Methoxysalicylaldehyde
5-Methoxysalicylaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 248
Melting Pt: 51…52 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9
EINECS: 211-589-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR