(±)-Mercaptosuccinic acid

Supplier: Molekula
Warning

Synonyms: (±)-Mercapto-1,4-butanedioic acid, 2-Mercaptosuccinic acid, (±)-Thiomalic acid, DL-Mercaptosuccinic acid

18131235-100G
MOLE18131235-100GEA 136 GBP
MOLE18131235-100G
(±)-Mercaptosuccinic acid
(±)-Mercaptosuccinic acid
Formula: C₄H₆O₄S
MW: 150.16 g/mol
Boiling Pt: 322
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004860
CAS Number: 70-49-5
EINECS: 200-736-4
Merck Index: 12,09479

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR