Pyridine-2,6-dicarboxylic acid

Supplier: Molekula
Warning

Synonyms: 2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA

14574395
MOLE14574395EA 0 GBP
MOLE14574395
Pyridine-2,6-dicarboxylic acid
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
Melting Pt: 327
Flash Pt: 188
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR