5-Bromopyrimidine

Supplier: Molekula
Warning

12306755-25G
MOLE12306755-25GEA 109 GBP
MOLE12306755-25G
5-Bromopyrimidine
5-Bromopyrimidine
Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 168 °C (727 mmHg)
Melting Pt: 75…76 °C
MDL Number: MFCD00006117
CAS Number: 4595-59-9
EINECS: 224-992-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR