L(+)-Ornithine hydrochloride

Supplier: Molekula

Synonyms: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

11702720-1KG
MOLE11702720-1KGEA 470 GBP
MOLE11702720-1KG
L(+)-Ornithine hydrochloride
L(+)-Ornithine hydrochloride
Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR