VITREX® Ultrasafe Safety Lancet
Supplier: Vitrex Medical A/S
397413
MODU397413EA 15.5 GBP
MODU397413
VITREX® Ultrasafe Safety Lancet
Blood Lancets
Vitrex Ultrasafe lancet is intended for blood glucose and other in vitro diagnostic tests.
  • Ergonomically optimised for better handling
  • High quality silicone coated needle; less pain and smooth penetration
  • Easy targeting of puncture site
  • Shelf life of five years
Health workers worldwide face accidental needle-stick injury while caring for patients, putting themselves at risk to bloodborne illnesses including the AIDS virus and Hepatitis.