Powered air respirator gaskets, Versaflo™
Supplier: 3M
M441
MMMAM441EA 45.5 GBP
MMMAM441
Powered air respirator gaskets, Versaflo™
Respirators Respiratory Systems
The Versaflo™ Jaw Gasket M-441 is a replacement jaw gasket for use with 3M™ Versaflo™ M-400 Headtop.
  • Spare or replacement part
  • Designed for use with 3M™ Versaflo™ M-400 Headtop
Prolong the service life of equipment with 3M™ Versaflo™ Jaw Gasket M-441. This jaw gasket is designed to replace worn or damaged parts in 3M™ Versaflo™ M-400 Headtop.