Filtration units, Aircare™
Supplier: 3M
ACU-04 ACU-03
MMMAACU-04EA 1170 GBP
MMMAACU-04 MMMAACU-03
Filtration units, Aircare™
Respirators Respiratory Systems
Aircare™ Filtration Units provide high-quality air from an external source. Removing water and oil droplets, solid particles, and odors, as well as condensation and contaminants from facility supplied compressed air, air is filtered for up to three users operating any 3M™ Supplied Air Respirator Systems.
  • Filter facility supplied compressed air
  • Can be used with any 3M™ Supplied Air Respirator System
  • Compact and lightweight designs
  • Quickly assembled and serviced
  • Available in wall-mounted or freestanding formats
Get high quality air from an externalMore Product Information