Spray adhesive, Super 77™
Supplier: 3M
77
MMMA77EA 28.2 GBP
MMMA77
Spray adhesive, Super 77™
Adhesives
This is an extremely versatile, fast-drying, and transparent spray adhesive that bonds a wide range of lightweight materials. The permanent bond provides fast results to keep projects moving ahead. The spray formula is easy to dispense and apply directly onto surfaces and materials where needed.
  • The low soak-in of the adhesive eliminates clamping and dry time
  • Repositionable while providing long-lasting strength
  • Soft, non-dimpling glue lines
  • Allows for a bond time of 15 seconds to 30 minutes
  • Typical applications include general assembly, temporary holding,
More Product Information