Scotch-Brite™ 3000 Diamond Floor Brush
Supplier: 3M
7100262371
MMMA7100262371EA 645 GBP
MMMA7100262371
Scotch-Brite™ 3000 Diamond Floor Brush
Wipes Cleaning Kits and Pads
Scotch-Brite™ Diamond floor brush 3000 series is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one step. Brush 3000 is a finer grit brush intended for daily use on floors where medium to high gloss is desired.
  • Provides both cleaning and gloss recovery in one step
  • Conformable, flexible diamond bristles reach low spots in floors
  • Longer life and durability when compared to traditional and diamond floor pads
  • A more sustainable solution as they can be used with just water to clean and maintain floors
  • Longer brush
More Product Information