Face shields, G500
Supplier: 3M
G5V5CH51 5V5F1H51
MMMAG5V5CH51EA 99.9 GBP
MMMAG5V5CH51 MMMA5V5F1H51
Face shields, G500
Face Shields
The G500 headgear provides a versatile and comfortable solution for wearers who require both face and hearing protection.
  • Headgear provides high impact resistance and a wide field of view
  • With ventilation in the front cover for increased comfort
  • Ratchet harness with ergonomically designed neck support and adjustment dial (3 positions for height adjustment, 8 positions for head size adjustment
  • Available with scratch and fog resistant PC visor for industrial applications or with stainless steel mesh visor for forestry applications
The special designMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable