Safety goggles, 2891
Supplier: 3M
2891-SG 2891S-SG
MMMA2891-SGEA 24.5 GBP
MMMA2891-SG MMMA2891S-SG
Safety goggles, 2891
Goggles
The 2891 series Safety Goggles are high-wrap, clear polycarbonate safety goggles with Scotchgard™ anti-fog/anti-scratch coating for superior protection.
  • Modern, slim design safety goggles, provide the wearer with an excellent field of vision
  • Anti-fog coating is effective even after multiple washes, disinfecting by soaking in diluted bleach, or using alcohol wipes
  • Wide, nylon elastic strap for simple adjustment and fit
  • Clear lens for excellent protection against Ultraviolet (UV) radiation
  • Strong polycarbonate lens for impact resistance
  • Lens marking: 2C-1.2 3
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable