Sampling valves
Supplier: Merck
MXPESP14N MXPESP18N MXPESPTCL MXPESP50R MXPESP5CN MXPESP5VT YY20040H0
MILXMXPESP14NEA 0 GBP
MILXMXPESP14N MILXMXPESP18N MILXMXPESPTCL MILXMXPESP50R MILXMXPESP5CN MILXMXPESP5VT MILXYY20040H0
Sampling valves
Valves Injection Valves
The sampling valves are used for sterile sampling of water or other final products in the pharmaceutical industry.
  • Collect samples mid-stream and filter through a MicropreSure unit, or sample into sterile sampling bags or a graduated cylinder
  • Samples are kept in sanitary condition for immediate sampling with alcohol or hydrogen peroxide injections
Materials: stem and body: 316L stainless steel or ECTFE body in ECTFE-type units, valve cap: ABS or PP, O-ring and plug seal: silicone or fluoroelastomer.
Certifications: Meets the FDA indirect foodMore Product Information