MultiScreen® Multiscreen Well Culture Trays
Supplier: Merck
MAMCS0110 MAMCS9610
MILFMAMCS0110EA 0 GBP
MILFMAMCS0110 MILFMAMCS9610
MultiScreen® Multiscreen Well Culture Trays
Culture Dishes Cell Culture Dishes
HTS filter plates: Multiscreen HTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics. Millipore's new Multiscreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensitiesMore Product Information