Chemicon® ReBlot Plus Kits

Supplier: Merck

2500-S 2500
MILF2500-SEA 104 GBP
MILF2500-S MILF2500
Chemicon® ReBlot Plus Kits
Electrophoresis Reagents Western Blotting Reagents

Western blotting is a commonly used technique for studying protein function and localisation. Typically, protein samples are electrophoresed on SDS-page and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid based technologies, which allow reuse of southern and northern blots, it has been difficult to reuse western blots.


  • Detection method: Chemiluminescence
  • Mild antibody stripping solution (10X) - (1 container, 50 ml)
  • Strong antibody stripping solution (10X) - (1 container, 50 ml)


The MILLIPORE re-blot plus western blot recycling kit is effective for removal of antibodies from western blots that have been developed with chemiluminescence or radioactive iodine or other isotopes. It is not recommended for stripping colorimetric substrates (TMB, DAB, 4-chloronapthol, and more.), as it is not possible to effectively remove substrates that precipitate at the reaction site.


The re-blot western blot recycling kit should be used only for qualitative purposes until it has been established by comparative blot analysis that stripping does not quantitatively affect a given antigen.


Caution: Note: To prevent reagent degradation secure the cap tightly upon storage. Avoid extended exposure to air.

Order Now

SPECIFICATIONS

Environmentally Preferable
Storage conditions Kit components should be stored at 4 °C upon arrival. Product is stable for 3 - 6 months after receipt. If antibody stripping solution crystallises upon storage, it may be re-dissolved with gentle warming at 37 °C before use.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR