96-Well Autosampler Vial Plate, with Inserts
Supplier: MICROSOLV
95096-710-D
MICS95096-710-DEA 261 GBP
MICS95096-710-D
96-Well Autosampler Vial Plate, with Inserts
Microplates Deep Well Microplates
96 well plates with glass inserts.
  • High grade, pure silicone rubber with PTFE coating
  • Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass
  • Chemically inert
The inserts are removable and the plates can be reused.