Detection window maker for capillary electrophoresis
Supplier: MICROSOLV
07201-S 07204-S 07207-S
MICS07201-SEA 241 GBP
MICS07201-S MICS07204-S MICS07207-S
Detection window maker for capillary electrophoresis
Electrophoresis Systems Capillary Electrophoresis Systems
Creates a perfect detection window. Removes the polyimide coating from 365 µm OD capillaries for detection by UV and LIF.
  • Easy to use for all capillaries for CE
  • Takes 10 seconds to make a perfect window
The Window Maker™ is designed to produce enough heat to burn off the polyimide but not damage the glass in any way. Can be used to remove 1 to 2 mm of the polyimide from the inlet and outlet tips of the capillary.
Delivery information: Supplied with a standard 2 mm module for burning the capillary. Other size heating modules are available as accessories.
Order Now
SPECIFICATIONS
Environmentally Preferable