Masterflex® Ismatec® Microbore Pump Tubing, FDA-Compliant Viton®, Avantor®
Supplier: Avantor Fluid Handling
MFLX97632-18EA 158 GBP
MFLX97632-18 MFLX97632-22 MFLX97632-24 MFLX97632-26 MFLX97632-28 MFLX97632-30 MFLX97632-32 MFLX97632-34 MFLX97632-36 MFLX97632-38 MFLX97632-40 MFLX97632-42 MFLX97632-44 MFLX97632-46 MFLX97632-48
Masterflex® Ismatec® Microbore Pump Tubing, FDA-Compliant Viton®, Avantor®
Tubing Peristaltic Pump Tubing
Excellent chemical resistance.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Viton® tubing has excellent chemical resistance, especially to acids and alkalis, making it an excellent choice for demanding applications such as acid and solvent transfer. ItMore Product Information