Masterflex® I/P® Precision Brushless Process Pumps, Avantor®
Supplier: Avantor Fluid Handling
MFLX77963-10EA 5170 GBP
MFLX77963-10 MFLX77963-10-EU MFLX77963-20-EU MFLX77963-10-UK MFLX77963-20-UK
Masterflex® I/P® Precision Brushless Process Pumps, Avantor®
Pumps Peristaltic Pumps
Powerful, low-maintenance drives are versatile and cost-effective.
  • Digital interface with LED display shows percent speed with 1% resolution
  • Speed control via three-turn potentiometer for quick, easy adjustments
  • Easily control start/stop and reverse features via sealed keypad
  • Contructed of non-chip painted steel and aluminum IP55-rated housing
  • Brushless, maintenance-free motor offers ±0.25% speed control accuracy
I/P® brushless process drives offer heavy-duty performance for your demanding process applications. This model features robust 1⁄3-hpMore Product Information