Masterflex® I/P® High-Performance Pump Head Replacement Parts, Avantor®
Supplier: Avantor Fluid Handling
MFLX77600-69EA 1110 GBP
MFLX77600-69
Masterflex® I/P® High-Performance Pump Head Replacement Parts, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Get the most our of your Masterflex® I/P® High-Performance Pump Head with these replacement parts.
  • Stainless steel (SS) rotor / roller for greater durability and chemical resistance
Replacement parts to keep your pump head operating even longer.