Masterflex® Pump Leak Detector Sensors, Avantor®
Supplier: Avantor Fluid Handling
MFLX77097-10EA 396 GBP
MFLX77097-10 MFLX77097-11 MFLX77097-12 MFLX77097-13 MFLX77097-22 MFLX77097-26 MFLX77097-28 MFLX77097-24
Masterflex® Pump Leak Detector Sensors, Avantor®
Leak Detection Systems Liquid Leak Detection Systems
Sensors shut down the pump drive in less than 1 second in the event of fluid leak or tubing failure.
  • Enhance safety of your Masterflex pump with leak detection and automatic shut down
  • Options available for mounting and connecting sensors to match your equipment and application
Leak detectors may be used with pumps that have an Open Head Sensor (OHS) input on the front panel or via the START/STOP input on the rear DB25 Interface Connection. Connection to START/STOP input requires use of Interface Adaptor (77097-24) and Extension Cable (77097-26). TheMore Product Information