Masterflex® Large-Hose Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
75800-62
MFLX75800-62EA 9450 GBP
MFLX75800-62
Masterflex® Large-Hose Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Multiple designs—either dry pump cavity or lubricant filled—to best fit your system requirements.
  • Complete isolation of pumped fluids
  • Self-priming and runs dry
  • Wide range of pump configurations and flows
  • No cups, packing, or seals for low maintenance
  • Will not break up delicate emulsions or create excessive frothing
The versatile VECTOR peristaltic pump series gives you pumping options you can’t get from standard positive displacement or centrifugal industrial pumps. The VECTOR pump is designed to handle the high-viscosity fluids commonly found inMore Product Information