Masterflex® Low-Flow Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
73160-00 73160-05 73160-10 73160-20 73160-25 73160-15
MFLX73160-00EA 314 GBP
MFLX73160-00 MFLX73160-05 MFLX73160-10 MFLX73160-20 MFLX73160-25 MFLX73160-15
Masterflex® Low-Flow Peristaltic Pumps, Avantor®
Pumps Microfluidic Pumps
Variable-flow reversible peristaltic pump.
  • Flow ranges from 0.03 to 4 ml/min
  • Run dry without damage to pump
These cost-effective peristaltic pumps provide variable flow and exceptional life performance in a compact package. Select from three different motor speeds, and three different tubing sizes to achieve flow rates from 0.03 to 4 ml/min.
Order power supply separately. Wall mount power supply, 115 VAC/12 VDC with US standard plug sold separately as part number 73160-50. Wall mount power supply, 230 VAC/12 VDC with EU plug sold separately as partMore Product Information