Masterflex® Single-Use Bag Port Disc Flat Plate, Avantor®
Supplier: Avantor Fluid Handling
MFLX40331-16EA 35.3 GBP
MFLX40331-16 MFLX40331-17 MFLX40331-18 MFLX40331-19 MFLX40331-20 MFLX40331-21 MFLX40331-22 MFLX40331-23 MFLX40331-24 MFLX40331-25 MFLX40331-26 MFLX40331-27
Masterflex® Single-Use Bag Port Disc Flat Plate, Avantor®
Fittings and Connectors
Flexible, smooth flat base.
  • Barb sizes range from 1/8" to 1"
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.