Masterflex® Barbed Panel-Mount Micro Flow Male Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
MFLX40263-00EA 28.8 GBP
MFLX40263-00 MFLX40263-01 MFLX40263-02 MFLX40263-03 MFLX40263-04 MFLX40263-05 MFLX40263-06 MFLX40263-07 MFLX40263-08 MFLX40263-09 MFLX40263-10 MFLX40263-11 MFLX40263-12 MFLX40263-13 MFLX40263-14 MFLX40263-15
Masterflex® Barbed Panel-Mount Micro Flow Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination.
  • Aseptic connection and disconnection
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
  • A complete coupler requires a male and female fitting with same flow size (order separately). Compatible with Female Micro Quick-Disconnect fittings.
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass throughMore Product Information