Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Acrylic, Avantor®
Supplier: Avantor Fluid Handling
32477-00 32477-02 32477-08 32477-16 32477-78
MFLX32477-04EA 169 GBP
MFLX32477-04 MFLX32477-06 MFLX32477-10 MFLX32477-12 MFLX32477-14 MFLX32477-18 MFLX32477-20 MFLX32477-22 MFLX32477-28 MFLX32477-30 MFLX32477-80 MFLX32477-82 MFLX32477-00 MFLX32477-02 MFLX32477-08 MFLX32477-16 MFLX32477-78
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Acrylic, Avantor®
Flow Meters
Read measurements in an unobstructed 360° view.
  • Easy-to-read dual scales (GPM/LPM)
  • Extremely durable—machined from high-quality acrylic rod stock
  • One-piece, in-line body occupies less space for a clean installation
These meters are designed for use in more aggressive process environments. In addition to the machined acrylic bodies, the design utilizes stainless steel float guides. The combination is a meter suitable for high-vibration, high-fluctuation process applications. Some versions include polypropylene end fittings reinforced with aluminumMore Product Information