Masterflex® Direct-Read Variable-Area Flowmeters for Water, Panel-Mount, Avantor®
Supplier: Avantor Fluid Handling
32471-00 32471-01 32471-03
MFLX32471-02EA 192 GBP
MFLX32471-02 MFLX32471-04 MFLX32471-00 MFLX32471-01 MFLX32471-03
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Panel-Mount, Avantor®
Flow Meters
Get better results than acrylic.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get a flow rate
  • Use with water for laboratory and industrial applications
These panel-mount flowmeters feature rotatable 90° elbow adapters for panel-mount installation. Installation is easy—simply drill two holes in your panel, insert meter fittings through the holes, secure with lock nuts (included), and rotate scale for easy viewing.