Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Water, 150-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
32000-28 32000-38 32001-28 32001-32 32001-36 32001-38 32002-34 32002-36 32003-30 32003-32 32003-34 32004-34 32004-36 32005-30 32005-32
MFLX32000-34EA 185 GBP
MFLX32000-34 MFLX32001-30 MFLX32001-34 MFLX32002-28 MFLX32002-30 MFLX32002-38 MFLX32003-28 MFLX32003-36 MFLX32003-38 MFLX32004-38 MFLX32005-28 MFLX32005-34 MFLX32005-36 MFLX32005-38 MFLX32030-40 MFLX32032-40 MFLX32033-40 MFLX32033-42 MFLX32035-40 MFLX32035-42 MFLX32000-28 MFLX32000-38 MFLX32001-28 MFLX32001-32 MFLX32001-36 MFLX32001-38 MFLX32002-34 MFLX32002-36 MFLX32003-30 MFLX32003-32 MFLX32003-34 MFLX32004-34 MFLX32004-36 MFLX32005-30 MFLX32005-32
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Water, 150-mm Scale, Avantor®
Flow Meters
No more eye strain—front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
More Product Information