Masterflex® L/S® Single-Use High-Performance Sanitary Tubing Assembly, Gamma Irradiated, Avantor®
Supplier: Avantor Fluid Handling
MFLX31201-38EA 408 GBP
MFLX31201-38 MFLX31201-39 MFLX31201-42
Masterflex® L/S® Single-Use High-Performance Sanitary Tubing Assembly, Gamma Irradiated, Avantor®
Tubing Transfer Tubing
Easy installation into your high-purity sanitary setups.
  • Obtain a contaminant-free sample from a WFI use point
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application