Masterflex® Tuf-Steel® Sanitary Tri-Clamp® Gaskets, Avantor®
Supplier: Avantor Fluid Handling
30547-04 30547-08 30547-10
MFLX30547-00EA 65.8 GBP
MFLX30547-00 MFLX30547-02 MFLX30547-06 MFLX30547-04 MFLX30547-08 MFLX30547-10
Masterflex® Tuf-Steel® Sanitary Tri-Clamp® Gaskets, Avantor®
Seals Gaskets
The benefits of PTFE and 316L stainless steel in one gasket!
  • Ideal for SIP and WFI applications
  • No cold flow or creep
  • Low extractables and leachables
The Tuf-Steel gasket combines the outstanding chemical resistance of a fluoropolymer with the rugged durability of stainless steel. Made of a unique blend of PTFE and 316L passivated stainless steel, this gasket eliminates the potential for gouging and forms a leakproof seal. It contains no pigments and has a pharmaceutical-grade inner diameter finish. Virtually impervious to steam, they are anMore Product Information