Masterflex® Turbine Flowmeter Single-Use Flow Tubes, Avantor®
Supplier: Avantor Fluid Handling
MFLX24507-14EA 762 GBP
MFLX24507-14 MFLX24507-15 MFLX24507-11 MFLX24507-12 MFLX24507-13 MFLX24507-36
Masterflex® Turbine Flowmeter Single-Use Flow Tubes, Avantor®
Fluid Sensors
Build a complete single-use flow sensor system customizable to your application.
  • Modular interchangeable flow sensor systems with single-use design
  • Accurate 1% of reading measurement at low flow
  • Low operating cost disposable flow tubes for optimal contamination-free production
  • Ideal for single-use applications while also suitable for long-term measurement
  • Flow tubes and turbine sensors are produced and tested under cleanroom conditions
The Masterflex® single-use flow sensor system is comprised of four components. Select the flow sensor, flow tubes,More Product Information