FMI Temperature-Controlled Piston Pump Heads
Supplier: Avantor Fluid Handling
07107-00 07107-05
MFLX07107-02EA 1040 GBP
MFLX07107-02 MFLX07107-04 MFLX07107-07 MFLX07107-09 MFLX07107-00 MFLX07107-05
FMI Temperature-Controlled Piston Pump Heads
Pumps Pump Heads Piston Pump Heads
Pump viscous fluids or maintain process fluid temperatures with temperature-controlled pump head.
  • Operating temperature up to 350 °F
  • Handle pressures up to 100 psig
  • Wetted materials of ceramic, PTFE, and 316 SS
Suitable for applications to maintain process fluid temperatures or pump viscous solutions (such as waxes, pastes or cosmetics). Run wash fluids, gases or steam through gland ports to provide a barrier between process fluids and the seal area (atmosphere). Using this barrier maintains temperatures and pressures required of slurries,More Product Information