Masterflex® Correlated Variable Area Flowmeters, PTFE Housing, 65-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
03216-50 03216-55 03216-57 03216-62 03216-65 03216-80 03266-55 03266-77 03266-80 32044-50 32044-55 32044-62 32044-67 32044-70 32044-80 32044-88
MFLX03216-52EA 307 GBP
MFLX03216-52 MFLX03216-60 MFLX03216-67 MFLX03216-70 MFLX03216-75 MFLX03216-77 MFLX03216-82 MFLX03216-85 MFLX03216-87 MFLX03266-65 MFLX03266-67 MFLX03266-82 MFLX32044-52 MFLX32044-57 MFLX32044-65 MFLX32044-77 MFLX32044-82 MFLX32044-87 MFLX32044-90 MFLX03216-50 MFLX03216-55 MFLX03216-57 MFLX03216-62 MFLX03216-65 MFLX03216-80 MFLX03266-55 MFLX03266-77 MFLX03266-80 MFLX32044-50 MFLX32044-55 MFLX32044-62 MFLX32044-67 MFLX32044-70 MFLX32044-80 MFLX32044-88
Masterflex® Correlated Variable Area Flowmeters, PTFE Housing, 65-mm Scale, Avantor®
Flow Meters
Designed for high-purity applications with flexibility for a broad array of gases.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
The substitution of metal fittings with PTFE eliminates a potential contamination source for high-purity applications. The glass tube utilizes a fused ceramic scale for a precise, permanent measuring guide. In addition, a vertical-tangential locatorMore Product Information