Sterilisers, SFplus
Supplier: MEMMERT
SF110PLUS SF160PLUS SF450PLUS SF260PLUS SF30PLUS SF55PLUS SF750PLUS SF75PLUS
MEMMSF110PLUSEA 4140 GBP
MEMMSF110PLUS MEMMSF160PLUS MEMMSF450PLUS MEMMSF260PLUS MEMMSF30PLUS MEMMSF55PLUS MEMMSF750PLUS MEMMSF75PLUS
Sterilisers, SFplus
Sterilisers Hot Air Sterilisers
These sterilisers comply with all relevant standards and regulations for medical products, and guarantee reliable program sequences, irrespective of the load and chamber volume. With the SetpointWAIT function validation of this hot air steriliser is possible anytime. The SetpointWAIT function ensures that the program only continues when the set point temperature has been reached. Precisely maintaining the sterilisation time and thus completely killing off even highly resistant microorganisms is, therefore, guaranteed at all times, irrespective of the sterilisation load and chamber volume.More Product Information