Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 3 µm
Supplier: Avantor
MDKB-3-1546EA 1740 GBP
MDKB-3-1546 MDKB-3-1503 MDKB-3-1502 MDKB-3-1046 MDKB-3-1003 MDKB-3-1002 MDKB-3-0546 MDKB-3-0503 MDKB-3-0502
Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 3 µm
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (3 µm and 5 µm ) and complementary phases
  • Analytical IDs in a
More Product Information