Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 5 µm
Supplier: Avantor
MDKA-5-2546UEA 1490 GBP
MDKA-5-2546U MDKA-5-2503U MDKA-5-2502U MDKA-5-1546U MDKA-5-1503U MDKA-5-1502U MDKA-5-1046U MDKA-5-1003U MDKA-5-1002U MDKA-5-0546U MDKA-5-0503U MDKA-5-0502U
Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 5 µm
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 µm and 5 µm ) and complementary phases
  • Analytical IDs in
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable