Cellulose MN 2100 adsorbents for column chromatography

Supplier: MACHEREY-NAGEL

Synonyms: Microcrystallinecellulose

815060.1 815060.5
MANA815060.1EA 53 GBP
MANA815060.1 MANA815060.5
Cellulose MN 2100 adsorbents for column chromatography
Cellulose

Native fibrous cellulose, purified grade (washed with different eluents) with average degree of polymerization 620–680.


  • For separation of hydrophilic substances
  • Fiber length (85%): 20 to 75 μm
  • Specific surface according to Blaine ~5500 cm²/g

Formula: (C₁₂H₂₂O₁₁)n
MW: 342.3 g/mol
Melting Pt: 260…270 °C
Density: 1.27…1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Order Now

Specification Test Results

Residue on Ignition <10000 ppm
Fe <2 ppm
Cu <1 ppm
P <2 ppm
Dichloromethane Extract <0.15%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR