Nessler reagent, tablets

Supplier: Lovibond

Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

511261BT 511231BT 511230BT 511251BT 511250BT
LOVI511261BTEA 137 GBP
LOVI511261BT LOVI511231BT LOVI511230BT LOVI511251BT LOVI511250BT
Nessler reagent, tablets
Nessler's reagent
MDL Number: MFCD00049656
CAS Number: 7783-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR