Scientific Ultra-Low-Temperature Freezer, Mediline SUFsg 5001
Supplier: Liebherr
SUFSG5001 SUFSG3501
LIEBSUFSG5001EA 0 GBP
LIEBSUFSG5001 LIEBSUFSG3501
Scientific Ultra-Low-Temperature Freezer, Mediline SUFsg 5001
Refrigerators
Liebherr appliances are optimised for maximum energy efficiency and low operating costs. We only use the natural and environmentally-friendly refrigerants R 290. When used with highly efficient compressors, these are extraordinarily efficient and, with their low global warming potential, also future-proof.
  • Data logger is integrated, USB interface
  • Includes power failure alarm and warning signal (optical and acoustical)
  • Aluminium slimline door handle with integrated opening mechanism
  • Replaceable door seals
Liebherr guarantees the outstanding quality ofMore Product Information