Seal with Bore for Screw Caps, Silicone with PTFE Washer
Supplier: Lenz Laborglas GmbH & CO.KG
01331810 01331406 01332508 01332510 01332512 01333210 01333212 01333214 01333216 01333218 01334526 01334532 01331808 01331806
201-0251EA 8.5 GBP
201-0251 201-0250 LENZ01332508 LENZ01332510 201-0239 LENZ01333210 LENZ01333212 LENZ01333214 LENZ01333216 201-0240 LENZ01334526 LENZ01334532 LENZ01331808 LENZ01331806
Seal with Bore for Screw Caps, Silicone with PTFE Washer
Seals Sealing Rings
Silicone rubber sealing rings with PTFE washers.
  • Temperature resistant from −200 to +250 °C
  • Excellent chemical resistance
  • Easy handling
For threaded connection caps with central bore and GL thread.
Order Now
SPECIFICATIONS
Environmentally Preferable