Portable cart
Supplier: LABCONCO
8025000
LBCP8025000EA 0 GBP
LBCP8025000
Portable cart
Trolleys
Safely transports heavy instruments and bulky equipment. Holds up to 181 kg. Welded frame is constructed of rugged, 18-gauge glacier white epoxy-coated 2,5 cm square tubular steel.
  • Shelves are chemical- and heat-resistant phenolic board with high-pressure melamine laminate surface
  • Cart features four 10,2 cm diameter casters (two with toe locks) and two handles with ergonomic extruded rubber grips