Freeze dryer accessories, FreeZone®
Supplier: LABCONCO
7815302 7369600 7379000 7379001 7369700 7370001 7370000
LBCP7815302EA 665 GBP
LBCP7815302 LBCP7369600 LBCP7379000 LBCP7379001 LBCP7369700 LBCP7370001 LBCP7370000
Freeze dryer accessories, FreeZone®
Freeze Dryers
Racks and supports hold test tubes, flasks and microwell plates for pre-freezing and freeze drying samples; clear canister holds chemical traps.
  • Racks feature sturdy steel or aluminium construction; Canister is acrylic
  • Designed to work with FreeZone® freeze dryers and drying chambers
  • Easily cleaned
The round test tube rack holds up to (104) 12 to 13 mm tubes for freeze drying.
Made of type 304 stainless steel and includes bottom. It is ideal for use with round Product Shelves (heated or non-heated). The round test tube/microcentrifuge tube rackMore Product Information