Heating and cooling process thermostats, Integral XT
Supplier: LAUDA
L002866 L002868 L002683 L002681
LAUDL002866EA 0 GBP
LAUDL002866 LAUDL002868 LAUDL002683 LAUDL002681
Heating and cooling process thermostats, Integral XT
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
Process thermostats with integrated cooling system for dynamic temperature control within external circuits.
  • Coloured TFT display for simultaneous indication of actual and set values and graphic illustration of the temperature profile
  • Clear text menu navigation, six selectable languages DE, EN, FR, ES, IT and RU
  • Management of heat transfer liquids with stored properties
  • Easy input via cursor and soft keys. Additional Tmax key for overtemperature protection
  • SelfCheck Assistant for system diagnosis
  • Fully electronic continuous controller with PID action for
More Product Information
New Product
Order Now
SPECIFICATIONS
Model IN 150 XT IN 250 XTW IN 8 XTW IN 2560 XTW
Environmentally Preferable
Electrical 230 V, 50 Hz 230 V, 50 Hz 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz